siRNA Details
| Gene Symbol: Myadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183332.1 | NP_899161.1 | 3 | 830 | ||
| XM_006228035.2 | XP_006228097.1 | 3 | 1021 | |||
| GenBank mRNA | AY344060.1 | - | 3 | 830 | ||
| BC087054.1 | - | 2 | 741 | 7123822 | ||
| BC107558.1 | - | 2 | 842 | 7932126 | ||
| BC128719.1 | - | 3 | 987 | 7315920 | ||

Myadm
Genomic Map