siRNA Details
| Gene Symbol: Ddx24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199119.1 | NP_954550.1 | 5 | 1593 | ||
| XM_006240483.1 | XP_006240545.1 | 5 | 1796 | |||
| XM_006240484.2 | XP_006240546.1 | 5 | 2028 | |||
| GenBank mRNA | AY341876.1 | - | 5 | 1593 | ||
| BC097262.1 | - | 5 | 1580 | 7442976 | ||
| FQ212095.1 | - | 3 | 339 | |||
| Gene Symbol: LOC100909476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750226.3 | XP_003750274.3 | 2 | 922 | ||

Ddx24
Genomic Map