siRNA Details
Gene Symbol: Il18rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767060.1 | XP_008765282.1 | 3, 4 | 1046 | ||
GenBank mRNA | AJ550893.1 | - | 1, 2 | 63 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il18rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767060.1 | XP_008765282.1 | 3, 4 | 1046 | ||
GenBank mRNA | AJ550893.1 | - | 1, 2 | 63 |