siRNA Details
| Gene Symbol: Prkag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_184051.1 | NP_908940.1 | 2 | 202 | ||
| XM_006235949.1 | XP_006236011.1 | 6 | 870 | |||
| XM_006235950.1 | XP_006236012.1 | 6 | 867 | |||
| XM_006235951.1 | XP_006236013.1 | 6 | 870 | |||
| XM_006235952.1 | XP_006236014.1 | 6 | 1773 | |||
| XM_006235953.2 | XP_006236015.1 | 6 | 3479 | |||
| XM_006235955.2 | XP_006236017.1 | 4 | 1743 | |||
| XM_006235957.2 | XP_006236019.1 | 2 | 4506 | |||
| XM_008762653.1 | XP_008760875.1 | 3 | 754 | |||
| XM_008762654.1 | XP_008760876.1 | 2 | 4502 | |||
| GenBank mRNA | AY348865.1 | - | 2 | 202 | ||
| AY376854.1 | - | 1 | 55 | |||
| BC097267.1 | - | 4 | 449 | |||
| FQ223364.1 | - | 6 | 1460 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001170556, NM_145401, NM_001170555, BQ572978, AK165834, and others.

Prkag2
Genomic Map