siRNA Details
| Gene Symbol: Tmem159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194354.2 | NP_919335.2 | 5 | 457 | ||
| XM_006230138.2 | XP_006230200.1 | 4 | 550 | |||
| XM_006230139.2 | XP_006230201.1 | 5 | 595 | |||
| XM_008759784.1 | XP_008758006.1 | 4 | 1388 | |||
| XM_008759785.1 | XP_008758007.1 | 4 | 1383 | |||
| GenBank mRNA | AY368496.1 | - | 3 | 291 | ||
| BC076384.1 | - | 5 | 457 | 7107584 | ||

Tmem159
Genomic Map