siRNA Details
| Gene Symbol: Trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198051.1 | NP_932168.1 | 2 | 155 | ||
| GenBank mRNA | AF532978.1 | - | 2 | 155 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trmt11| Gene Symbol: Trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198051.1 | NP_932168.1 | 2 | 155 | ||
| GenBank mRNA | AF532978.1 | - | 2 | 155 | ||