siRNA Details
| Gene Symbol: Pcdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199503.1 | NP_955797.1 | 1 | 919 | ||
| GenBank mRNA | AB113384.1 | - | 1 | 919 | ||
| AY573971.1 | - | 1 | 959 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha1| Gene Symbol: Pcdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199503.1 | NP_955797.1 | 1 | 919 | ||
| GenBank mRNA | AB113384.1 | - | 1 | 919 | ||
| AY573971.1 | - | 1 | 959 | |||