siRNA Details
| Gene Symbol: Pcdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199504.1 | NP_955798.1 | 1 | 916 | ||
| GenBank mRNA | AB113385.1 | - | 1 | 916 | ||
| AY573976.1 | - | 1 | 916 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha2| Gene Symbol: Pcdha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199504.1 | NP_955798.1 | 1 | 916 | ||
| GenBank mRNA | AB113385.1 | - | 1 | 916 | ||
| AY573976.1 | - | 1 | 916 | |||