siRNA Details
Gene Symbol: Pcdhac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201422.1 | NP_958825.1 | 1 | 1613 | ||
GenBank mRNA | AB113398.1 | - | 1 | 1613 | ||
AY573985.1 | - | 1 | 1613 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
