siRNA Details
| Gene Symbol: Pcdhac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201422.1 | NP_958825.1 | 1 | 2410 | ||
| GenBank mRNA | AB113398.1 | - | 1 | 2410 | ||
| AY573985.1 | - | 1 | 2410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhac2| Gene Symbol: Pcdhac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201422.1 | NP_958825.1 | 1 | 2410 | ||
| GenBank mRNA | AB113398.1 | - | 1 | 2410 | ||
| AY573985.1 | - | 1 | 2410 | |||