siRNA Details
| Gene Symbol: Pcdha11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199486.2 | NP_955780.1 | 1 | 1137 | ||
| GenBank mRNA | AB113394.1 | - | 1 | 1137 | ||
| AY573973.1 | - | 1 | 1137 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdha11| Gene Symbol: Pcdha11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199486.2 | NP_955780.1 | 1 | 1137 | ||
| GenBank mRNA | AB113394.1 | - | 1 | 1137 | ||
| AY573973.1 | - | 1 | 1137 | |||