siRNA Details
| Gene Symbol: Ugt1a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201425.2 | NP_958828.1 | 1 | 207 | ||
| GenBank mRNA | AY435129.1 | - | 1 | 207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt1a9| Gene Symbol: Ugt1a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201425.2 | NP_958828.1 | 1 | 207 | ||
| GenBank mRNA | AY435129.1 | - | 1 | 207 | ||