siRNA Details
| Gene Symbol: E2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100778.1 | NP_001094248.1 | 3 | 534 | ||
| XM_008762363.1 | XP_008760585.1 | 3 | 531 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E2f1| Gene Symbol: E2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100778.1 | NP_001094248.1 | 3 | 534 | ||
| XM_008762363.1 | XP_008760585.1 | 3 | 531 | |||