siRNA Details
| Gene Symbol: aicda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100779.1 | NP_001094249.1 | 3, 4 | 514 | ||
| GenBank mRNA | AJ606071.2 | - | 2, 3 | 335 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

aicda| Gene Symbol: aicda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100779.1 | NP_001094249.1 | 3, 4 | 514 | ||
| GenBank mRNA | AJ606071.2 | - | 2, 3 | 335 | ||