siRNA Details
| Gene Symbol: Vkorc1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203338.1 | NP_976083.1 | 3 | 489 | ||
| GenBank mRNA | AY423048.1 | - | 3 | 489 | ||
| Gene Symbol: LOC103693015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766000.1 | XP_008764222.1 | 3 | 472 | ||
| XM_008766001.1 | XP_008764223.1 | 2 | 362 | |||

Vkorc1l1
Genomic Map