siRNA Details
| Gene Symbol: Mid1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206950.1 | NP_996833.1 | 2 | 911 | ||
| XM_006256672.2 | XP_006256734.1 | 2 | 731 | |||
| GenBank mRNA | AB354580.1 | - | 2 | 808 | ||
| BC061715.1 | - | 2 | 911 | 5598493 | ||

Mid1ip1
Genomic Map