siRNA Details
| Gene Symbol: Olr785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000603.1 | NP_001000603.1 | 1 | 671 | ||
| Gene Symbol: Olr781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000375.1 | NP_001000375.1 | 1 | 671 | ||

Olr785
Genomic Map