siRNA Details
| Gene Symbol: Olr784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000604.1 | NP_001000604.1 | 765 | |||
| Gene Symbol: LOC100909573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749538.1 | XP_003749586.1 | 1 | 765 | ||

Olr784
Genomic Map