siRNA Details
| Gene Symbol: Olr780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000606.1 | NP_001000606.1 | 1 | 857 | ||
| Gene Symbol: Olr779 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000917.1 | NP_001000917.1 | 1 | 857 | ||

Olr780
Genomic Map