siRNA Details
| Gene Symbol: Olr737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000616.1 | NP_001000616.1 | 1 | 384 | ||
| GenBank mRNA | X89698.1 | - | 1 | 19 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr737| Gene Symbol: Olr737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000616.1 | NP_001000616.1 | 1 | 384 | ||
| GenBank mRNA | X89698.1 | - | 1 | 19 | ||