siRNA Details
| Gene Symbol: Olr734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000617.1 | NP_001000617.1 | 1 | 400 | ||
| Gene Symbol: LOC103694849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773785.1 | XP_008772007.1 | 1 | 400 | ||
| Gene Symbol: Olr733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000618.1 | NP_001000618.1 | 1 | 400 | ||

Olr734
Genomic Map