siRNA Details
| Gene Symbol: USP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128610.2 | NP_001122082.1 | 12 | 2214 | ||
| NM_005154.4 | NP_005145.3 | 12 | 2074 | |||
| XM_006720761.3 | XP_006720824.1 | 13 | 2394 | |||
| XM_011522193.2 | XP_011520495.1 | 13 | 2231 | |||
| XM_017022721.1 | XP_016878210.1 | 13 | 2131 | |||
| XM_017022722.1 | XP_016878211.1 | 11 | 2054 | |||
| GenBank mRNA | AK292400.1 | - | 12 | 2069 | ||
| BC110590.2 | - | 12 | 2040 | 40027270 | ||
| BX537420.1 | - | 12 | 2069 | |||
| D29956.1 | - | 12 | 2188 | |||

USP8
Genomic Map