siRNA Details
| Gene Symbol: Olr727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000619.1 | NP_001000619.1 | 1 | 103 | ||
| Gene Symbol: Olr726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000620.2 | NP_001000620.2 | 1, 2 | 43 | ||
| Gene Symbol: Olr728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000363.1 | NP_001000363.1 | 1 | 100 | ||

Olr727
Genomic Map