siRNA Details
| Gene Symbol: Olr716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000622.1 | NP_001000622.1 | 2 | 784 | ||
| Gene Symbol: Olr712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000624.1 | NP_001000624.1 | 1 | 784 | ||
| Gene Symbol: Olr714 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000923.1 | NP_001000923.1 | 1 | 784 | ||

Olr716
Genomic Map