siRNA Details
| Gene Symbol: Olr654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000636.1 | NP_001000636.1 | 1 | 789 | ||
| Gene Symbol: Olr652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000638.1 | NP_001000638.1 | 789 | |||

Olr654
Genomic Map