siRNA Details
| Gene Symbol: Olr646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000642.1 | NP_001000642.1 | 1 | 674 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr646| Gene Symbol: Olr646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000642.1 | NP_001000642.1 | 1 | 674 | ||