siRNA Details
| Gene Symbol: Olr640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000644.1 | NP_001000644.1 | 1 | 511 | ||
| Gene Symbol: Olr635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000646.1 | NP_001000646.1 | 1 | 511 | ||

Olr640
Genomic Map