siRNA Details
Gene Symbol: Olr621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000652.1 | NP_001000652.1 | 511 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000652.1 | NP_001000652.1 | 511 |