siRNA Details
| Gene Symbol: Olr569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000664.1 | NP_001000664.1 | 1 | 319 | ||
| Gene Symbol: Olr575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000327.1 | NP_001000327.1 | 1 | 319 | ||

Olr569
Genomic Map