siRNA Details
| Gene Symbol: Olr566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000665.1 | NP_001000665.1 | 1 | 784 | ||
| Gene Symbol: Olr531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001051.1 | NP_001001051.1 | 1 | 784 | ||

Olr566
Genomic Map