siRNA Details
| Gene Symbol: Olr563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000666.1 | NP_001000666.1 | 1 | 661 | ||
| Gene Symbol: Olr531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001051.1 | NP_001001051.1 | 1 | 661 | ||

Olr563
Genomic Map