siRNA Details
| Gene Symbol: Olr557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000669.1 | NP_001000669.1 | 1 | 471 | ||
| Gene Symbol: Olr556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000929.1 | NP_001000929.1 | 1 | 372 | ||

Olr557
Genomic Map