siRNA Details
| Gene Symbol: Olr529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000674.1 | NP_001000674.1 | 1 | 266 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr529| Gene Symbol: Olr529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000674.1 | NP_001000674.1 | 1 | 266 | ||