siRNA Details
| Gene Symbol: Olr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000690.1 | NP_001000690.1 | 1 | 99 | ||
| Gene Symbol: Olr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000936.1 | NP_001000936.1 | 1 | 99 | ||
| Gene Symbol: Olr29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000691.1 | NP_001000691.1 | 1 | 99 | ||

Olr32
Genomic Map