siRNA Details
| Gene Symbol: Olr29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000691.1 | NP_001000691.1 | 1 | 946 | ||
| Gene Symbol: Olr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000690.1 | NP_001000690.1 | 1 | 946 | ||

Olr29
Genomic Map