siRNA Details
| Gene Symbol: Olr954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000703.1 | NP_001000703.1 | 775 | |||
| Gene Symbol: LOC100911571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765074.1 | XP_008763296.1 | 1 | 775 | ||
| Gene Symbol: LOC103692768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765076.1 | XP_008763298.1 | 1 | 775 | ||

Olr954
Genomic Map