siRNA Details
| Gene Symbol: Olr1479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000721.1 | NP_001000721.1 | 1 | 291 | ||
| Gene Symbol: Olr1472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000944.1 | NP_001000944.1 | 1 | 291 | ||

Olr1479
Genomic Map