siRNA Details
| Gene Symbol: Olr1471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000722.1 | NP_001000722.1 | 2 | 578 | ||
| XM_008767911.1 | XP_008766133.1 | 2 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1471| Gene Symbol: Olr1471 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000722.1 | NP_001000722.1 | 2 | 578 | ||
| XM_008767911.1 | XP_008766133.1 | 2 | 578 | |||