siRNA Details
| Gene Symbol: Olr1468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000724.1 | NP_001000724.1 | 1 | 784 | ||
| GenBank mRNA | AF091567.1 | - | 595 | |||
| M64392.1 | - | 1 | 784 | |||
| Gene Symbol: LOC103693363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767910.1 | XP_008766132.1 | 1 | 784 | ||

Olr1468
Genomic Map