siRNA Details
| Gene Symbol: Olr1468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000724.1 | NP_001000724.1 | 1 | 307 | ||
| GenBank mRNA | AF091567.1 | - | 118 | |||
| M64392.1 | - | 1 | 307 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1468| Gene Symbol: Olr1468 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000724.1 | NP_001000724.1 | 1 | 307 | ||
| GenBank mRNA | AF091567.1 | - | 118 | |||
| M64392.1 | - | 1 | 307 | |||