siRNA Details
| Gene Symbol: Olr1539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000729.1 | NP_001000729.1 | 443 | |||
| Gene Symbol: Olr1537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001352.1 | NP_001001352.1 | 1 | 443 | ||

Olr1539
Genomic Map