siRNA Details
| Gene Symbol: Olr213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000735.1 | NP_001000735.1 | 1 | 94 | ||
| Gene Symbol: Olr211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000196.1 | NP_001000196.1 | 1 | 94 | ||

Olr213
Genomic Map