siRNA Details
| Gene Symbol: Olr164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000737.1 | NP_001000737.1 | 1 | 121 | ||
| Gene Symbol: Olr170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000175.1 | NP_001000175.1 | 121 | |||

Olr164
Genomic Map