siRNA Details
| Gene Symbol: Olr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001283.1 | NP_001001283.1 | 1 | 43 | ||
| Gene Symbol: Olr139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000161.1 | NP_001000161.1 | 1 | 25 | ||
| Gene Symbol: Olr140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000162.1 | NP_001000162.1 | 1 | 25 | ||

Olr137
Genomic Map