siRNA Details
| Gene Symbol: Olr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000745.1 | NP_001000745.1 | 1 | 144 | ||
| Gene Symbol: Olr88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000746.1 | NP_001000746.1 | 1 | 141 | ||

Olr89
Genomic Map