siRNA Details
| Gene Symbol: SLC16A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271765.1 | NP_001258694.1 | 6 | 1553 | ||
| NM_004695.3 | NP_004686.1 | 6 | 1487 | |||
| XM_005257790.1 | XP_005257847.1 | 5 | 1429 | |||
| XM_017025293.1 | XP_016880782.1 | 6 | 1515 | |||
| GenBank mRNA | AK000416.1 | - | 6 | 1553 | ||
| AK092512.1 | - | 5 | 1596 | |||
| AK304163.1 | - | 5 | 1313 | |||
| BC009684.1 | - | 6 | 1468 | 3895881 | ||
| BC033611.1 | - | 5 | 1377 | 5218712 | ||
| U59299.1 | - | 4 | 1241 | |||

SLC16A5
Genomic Map