siRNA Details
| Gene Symbol: Olr331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000760.1 | NP_001000760.1 | 1 | 564 | ||
| Gene Symbol: Olr334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001041.1 | NP_001001041.1 | 1 | 555 | ||

Olr331
Genomic Map