siRNA Details
| Gene Symbol: Olr1459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000770.1 | NP_001000770.1 | 1 | 323 | ||
| Gene Symbol: Olr1460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000769.1 | NP_001000769.1 | 1 | 323 | ||

Olr1459
Genomic Map