siRNA Details
Gene Symbol: Olr1459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000770.1 | NP_001000770.1 | 1 | 471 |
Gene Symbol: Olr1461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000022.1 | NP_001000022.1 | 1 | 471 |
