siRNA Details
| Gene Symbol: Olr1424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000779.1 | NP_001000779.1 | 1 | 50 | ||
| Gene Symbol: LOC103690286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767749.1 | XP_008765971.1 | 1 | 50 | ||

Olr1424
Genomic Map